accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132388-60-4

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-[(triphenylmethyl)carbamoyl]butanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-4-(triphenylmethylcarbamoyl)butanoic acid
Molecular Weight (g/mol): 522.6
Molecular Formula: C32H30N2O5
InChi Key: MYOAIKMOWHPBQS-UHFFFAOYNA-N
SMILES: OC(=O)C(CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)