Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Benzyloxycarbonyl-Ndelta-trityl-L-glutamine, 98%, Thermo Scientific™

Catalog No. AAH6354903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6354903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6354903 Supplier Thermo Scientific Chemicals Supplier No. H6354903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132388-60-4
Molecular Formula C32H30N2O5
Molecular Weight (g/mol) 522.601
MDL Number MFCD00144845
InChI Key MYOAIKMOWHPBQS-NDEPHWFRSA-N
Synonym z-gln trt-oh, cbz-gln trt-oh, z-l-gln trt-oh, n-cbz-n'-trityl-l-glutamine, s-2-benzyloxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-benzyloxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem15365, 2s-2-benzyloxycarbonylamino-5-oxo-5-tritylamino pentanoic acid
PubChem CID 10962399
IUPAC Name (2S)-5-oxo-2-(phenylmethoxycarbonylamino)-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)NC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)O

Specifications

Quantity 1 g
Formula Weight 522.59
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Ndelta-trityl-L-glutamine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.