accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH(S)OOHO(S)O(R)HO(S)OH(S)OH(R)O(S)O(S)H3C(R)HO(R)OH(R)OHO

CAS RN 13241-33-3

IUPAC Name: (2S)-7-{[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: neohesperidin neohesperidin (8CI) hesperetin 7-neohesperidoside
Molecular Weight (g/mol): 610.57
Molecular Formula: C28H34O15
InChi Key: ARGKVCXINMKCAZ-UZRWAPQLSA-N
SMILES: COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)C=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results