Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neohesperidin, 97.8%, MP Biomedicals™

Catalog No. ICN15581380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15581380 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15581380 Supplier MP Biomedicals, Inc Supplier No. 0215581380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soluble in pyridine (20mg/mL-clear, yellow solution)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13241-33-3
Molecular Formula C28H34O15
Molecular Weight (g/mol) 610.565
MDL Number MFCD16661259
InChI Key ARGKVCXINMKCAZ-CIDMPNHLSA-N
Synonym hesperetin-7-o-neohesperidoside
PubChem CID 134129340
IUPAC Name (2S)-7-[(2S,3R,4S,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3S,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
SMILES CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=CC(=C4C(=O)CC(OC4=C3)C5=CC(=C(C=C5)OC)O)O)CO)O)O)O)O)O

Specifications

Color Tan
Quantity 100 mg
Formula Weight 610.6
Physical Form Powder
Chemical Name or Material Neohesperidin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.