accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132465-11-3

IUPAC Name: 3-phenylprop-2-en-1-yl 2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms: 3-phenylprop-2-en-1-yl 2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 321.33
Molecular Formula: C19H15NO4
InChi Key: XGHYFEJMJXGPGN-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C(C#N)C(=O)OCC=CC2=CC=CC=C2)C=C1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)