CAS RN 132499-82-2
IUPAC Name:
disodium propanedioate
Synonyms:
sodium malonate|malonic acid disodium salt|malonic acid, disodium salt|malonic acid, sodium salt|sodium malonate dibasic|propanedioic acid, disodium salt|propanedioic acid, sodium salt (1:2)
Molecular Weight (g/mol):
148.03
Molecular Formula:
C3H2Na2O4
InChi Key:
PRWXGRGLHYDWPS-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]C(=O)CC([O-])=O
1
–
1
of
1
results
1
Promotions Available
Malonic acid disodium salt hydrate, 99%
Malonic acid disodium salt hydrate, 99%
CAS: 132499-82-2 Molecular Formula: C3H2Na2O4 Molecular Weight (g/mol): 148.03 MDL Number: MFCD00149114 InChI Key: PRWXGRGLHYDWPS-UHFFFAOYSA-L IUPAC Name: disodium propanedioate SMILES: [Na+].[Na+].[O-]C(=O)CC([O-])=O