Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malonic acid disodium salt hydrate, 99%

Catalog No. AC413505000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413505000 500 g Plastic bottle
AC413500250 25 g Plastic bottle
AC413501000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC413505000 Supplier Thermo Scientific Chemicals Supplier No. 413505000
  • $488.50
    $415.65 / Each of 1

    Save $72.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 132499-82-2 | C3H2Na2O4 | 148.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132499-82-2
Molecular Formula C3H2Na2O4
Molecular Weight (g/mol) 148.03
MDL Number MFCD00149114
InChI Key PRWXGRGLHYDWPS-UHFFFAOYSA-L
IUPAC Name disodium propanedioate
SMILES [Na+].[Na+].[O-]C(=O)CC([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH2(COONa)2·xH2O
Quantity 500 g
Solubility Information Solubility in water: soluble
Formula Weight 148.03
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Malonic acid disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.