accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13250-12-9

IUPAC Name: butan-2-amine
Synonyms: sec-butylamine
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: BHRZNVHARXXAHW-UHFFFAOYNA-N
SMILES: CCC(C)N
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)