Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-Sec-Butylamine

Catalog No. 296643250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-664-3250MG 250 mg
29-664-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-664-3250MG Supplier Sigma Aldrich Supplier No. 296643250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-sec-Butylamine can be used:To prepare (R)-(-)-sec-butylbis(2-hydroxyethyl)amine, an intermediate for the synthesis of aminodiphosphine ligands.As a substrate to demonstrate the stereochemistry of Mitsunobu reaction in the synthesis of secondary amines.As a model compound in the study of singlet-triplet transitions in circular dichroism.

Specifications

CAS 13250-12-9
Density 0.72 g/mL at 25°C
Boiling Point 63°C
Molecular Formula C4H11N
Refractive Index n20/D 1.393
Linear Formula C2H5CH(NH2)CH3
MDL Number MFCD00064416
Quantity 250 mg
Synonym (R)-(-)-2-Aminobutane
Molecular Weight (g/mol) 73.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.