accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNOO

CAS RN 13269-19-7

IUPAC Name: 2-nitropyridin-3-amine
Synonyms: 2-nitropyridin-3-amine
Molecular Weight (g/mol): 139.11
Molecular Formula: C5H5N3O2
InChi Key: GZBKVUGZEAJYHH-UHFFFAOYSA-N
SMILES: NC1=CC=CN=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)