Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-nitropyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAL1961906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1961906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1961906 Supplier Thermo Scientific Chemicals Supplier No. L1961906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13269-19-7 | Molecular Formula: C5H5N3O2 | Molecular Weight: 139.114 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13269-19-7
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00044102
InChI Key GZBKVUGZEAJYHH-UHFFFAOYSA-N
Synonym 3-amino-2-nitropyridine, 2-nitro-3-pyridinamine, 2-nitro-3-aminopyridine, 3-pyridinamine, 2-nitro, 2-nitropyridin-3-amin, aminonitropyridine, amino nitro pyridine, pubchem2087, acmc-209bps, 2-nitro-3-pyridylamine
PubChem CID 83281
IUPAC Name 2-nitropyridin-3-amine
SMILES C1=CC(=C(N=C1)[N+](=O)[O-])N

Specifications

Melting Point 191°C to 193°C
Quantity 5 g
Beilstein 124467
Formula Weight 139.11
Percent Purity 97%
Chemical Name or Material 3-Amino-2-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 236-260-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.