accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1327-36-2

IUPAC Name: oxo[(oxoalumanyl)oxy]alumane; silanedione
Synonyms: zeolite
Molecular Weight (g/mol): 162.04
Molecular Formula: Al2O5Si
InChi Key: HNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILES: O=[Si]=O.O=[Al]O[Al]=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Material 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)