Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, Indicating, Reagents

Catalog No. 01337073
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-073 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-073 Supplier Reagents Holdings Llc Supplier No. C2300600500C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silica Gel, Indicating, Orange Expose to atmosphere only when put into use. Environmentally safe. Turns green when hydrated., Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1327-36-2
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.04
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
IUPAC Name oxo[(oxoalumanyl)oxy]alumane; silanedione
SMILES O=[Si]=O.O=[Al]O[Al]=O

Specifications

Physical Form Solid
Color Orange
Material Silica Gel
Quantity 500 g
Packaging Glass Bottle
Specific Gravity 1
Chemical Name or Material Silica Gel, Indicating, Orange Expose to atmosphere only when put into use. Environmentally safe. Turns green when hydrated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.