accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132715-69-6

FBrOHO

CAS RN 132715-69-6

IUPAC Name: 2-bromo-3-fluorobenzoic acid
Synonyms: 2-bromo-3-fluorobenzoic acid
Molecular Weight (g/mol): 219.01
Molecular Formula: C7H4BrFO2
InChi Key: KQRCBMPPEPNNDS-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C(F)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results