Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-fluorobenzoic Acid 96.0+%, TCI America™
SDP

Catalog No. B37225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37225G 5 g
B37221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37225G Supplier TCI America Supplier No. B37225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132715-69-6
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD01569398
InChI Key KQRCBMPPEPNNDS-UHFFFAOYSA-N
Synonym 3-fluoro-2-bromobenzoic acid, 2-bromo-3-fluorobenzoicacid, 2-bromo-3-fluoro-benzoic acid, buttpark 22\01-97, benzoic acid, 2-bromo-3-fluoro, pubchem1319, 2-bromo-3-fluorobenzoic, acmc-1c0re, intermediates-zcf02016, ksc174c5p
PubChem CID 302622
IUPAC Name 2-bromo-3-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)Br)C(=O)O

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.