accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOO

CAS RN 132732-60-6

IUPAC Name: 4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Synonyms: 4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular Weight (g/mol): 434.53
Molecular Formula: C27H30O5
InChi Key: PDGHLARNGSMEJE-UHFFFAOYNA-N
SMILES: OC1CC(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(COCC2=CC=CC=C2)O1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)