accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHNCl

CAS RN 13278-36-9

IUPAC Name: 2-[(3-chlorophenyl)amino]benzoic acid
Synonyms: 2-[(3-chlorophenyl)amino]benzoic acid
Molecular Weight (g/mol): 247.68
Molecular Formula: C13H10ClNO2
InChi Key: OVMWPVYEBVFZHM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1NC1=CC=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results