Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Chlorophenyl)anthranilic Acid 98.0+%, TCI America™
SDP

Catalog No. C206525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C206525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C206525G Supplier TCI America Supplier No. C206525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13278-36-9
Molecular Formula C13H10ClNO2
Molecular Weight (g/mol) 247.68
MDL Number MFCD01928172
InChI Key OVMWPVYEBVFZHM-UHFFFAOYSA-N
Synonym 2-[(3-Chlorophenyl)amino]benzoic Acid, 2-(3-Chloroanilino)benzoic Acid
PubChem CID 83293
IUPAC Name 2-[(3-chlorophenyl)amino]benzoic acid
SMILES OC(=O)C1=CC=CC=C1NC1=CC=CC(Cl)=C1

Specifications

Melting Point 171°C
Quantity 25 g
Formula Weight 247.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Chlorophenyl)anthranilic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.