accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNOO

CAS RN 13290-74-9

IUPAC Name: 1-chloro-2-methyl-4-nitrobenzene
Synonyms: 1-chloro-2-methyl-4-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: BGDCQZFFNFXYQC-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results