accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132927-09-4

IUPAC Name: 2-methoxy-4-(trifluoromethyl)benzaldehyde
Synonyms: 2-methoxy-4-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: KKESHHUPAFNMPH-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results