accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133-13-1

IUPAC Name: 1,3-diethyl 2-ethylpropanedioate
Synonyms: 1,3-diethyl 2-ethylpropanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: VQAZCUCWHIIFGE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC)C(=O)OCC
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)