Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ethylmalonate, 99%

Catalog No. AC114365000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114365000 500 g Glass bottle
AC114361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114365000 Supplier Thermo Scientific Chemicals Supplier No. 114365000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 133-13-1 | C9H16O4 | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-13-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00009167
InChI Key VQAZCUCWHIIFGE-UHFFFAOYSA-N
Synonym diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate
PubChem CID 8610
IUPAC Name diethyl 2-ethylpropanedioate
SMILES CCC(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Density 1.0000g/mL
Boiling Point 75°C to 77°C (5.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4150 to 1.417
Linear Formula C2H5O2CCH(C2H5)CO2C2H5
Quantity 500 g
Beilstein 02, 644
Specific Gravity 1
Solubility Information Solubility in water: immiscible
Formula Weight 188.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl ethylmalonate, 99%
Show More Show Less
EINECSNumber 205-093-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.