accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaNH2HOOO

CAS RN 133-15-3

IUPAC Name: calcium 4-amino-2-hydroxybenzoate
Synonyms: aminosalicylate calcium
Molecular Weight (g/mol): 192.21
Molecular Formula: C7H6CaNO3
InChi Key: JUBKIVBSIIADES-UHFFFAOYSA-M
SMILES: [Ca++].NC1=CC=C(C([O-])=O)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results