Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminosalicylic Acid Calcium Salt Heptahydrate 98.0+%, TCI America™
SDP

Catalog No. A0422500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0422500G 500 g
A042225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0422500G Supplier TCI America Supplier No. A0422500G

Chemical Identifiers

CAS 133-15-3
Molecular Formula C14H12CaN2O6
Molecular Weight (g/mol) 344.336
MDL Number MFCD00067430
InChI Key XDWVNCOPMIEDJK-UHFFFAOYSA-L
Synonym 4-Amino-2-hydroxybenzoic Acid Calcium Salt, Dicalcium Bis(4-aminosalicylate)
PubChem CID 102600765
IUPAC Name calcium;4-amino-2-hydroxybenzoate
SMILES C1=CC(=C(C=C1N)O)C(=O)[O-].C1=CC(=C(C=C1N)O)C(=O)[O-].[Ca+2]

Specifications

Quantity 500 g
Formula Weight 382.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminosalicylic Acid Calcium Salt Heptahydrate
RTECSNumber VO1470000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.