accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3ONNOHNBr

CAS RN 133040-77-4

IUPAC Name: 2-[2-(5-bromo-1H-indol-3-yl)ethyl]-3-[3-(propan-2-yloxy)phenyl]-3,4-dihydroquinazolin-4-one
Synonyms: 2-[2-(5-bromo-1H-indol-3-yl)ethyl]-3-(3-isopropoxyphenyl)quinazolin-4-one
Molecular Weight (g/mol): 502.41
Molecular Formula: C27H24BrN3O2
InChi Key: KUECXUACQOYKNB-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC(=C1)N1C(CCC2=CNC3=CC=C(Br)C=C23)=NC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results