accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOO(S)OHCH2

CAS RN 133095-74-6

IUPAC Name: (2S)-1-[(4-methylbenzenesulfonyl)oxy]but-3-en-2-ol
Synonyms: (2S)-1-[(4-methylbenzenesulfonyl)oxy]but-3-en-2-ol
Molecular Weight (g/mol): 242.29
Molecular Formula: C11H14O4S
InChi Key: YQSCDBZHHLIPOI-JTQLQIEISA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H](O)C=C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results