accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13311-84-7

IUPAC Name: 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Synonyms: flutamide niftolid flutamida FTA niftholide flutamidum eulexin cebatrol niftolide 氟他胺
Molecular Weight (g/mol): 276.22
Molecular Formula: C11H11F3N2O3
InChi Key: MKXKFYHWDHIYRV-UHFFFAOYSA-N
SMILES: CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results