accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOONH(R)(S)CH3

CAS RN 133180-01-5

IUPAC Name: (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(triphenylmethoxy)butanoic acid
Synonyms: (2S,3R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(triphenylmethoxy)butanoic acid
Molecular Weight (g/mol): 583.68
Molecular Formula: C38H33NO5
InChi Key: JARBLLDDSTVWSM-IJAHGLKVSA-N
SMILES: C[C@@H](OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)