accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13319-71-6

IUPAC Name: 2-bromo-6-methylphenol
Synonyms: 2-bromo-6-methylphenol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: YXZPTVOCJLCMRO-UHFFFAOYSA-N
SMILES: CC1=C(O)C(Br)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results