Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-o-cresol 94.0+%, TCI America™
SDP

Catalog No. B31265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31265G 5 g
B312625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31265G Supplier TCI America Supplier No. B31265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13319-71-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD09907876
InChI Key YXZPTVOCJLCMRO-UHFFFAOYSA-N
Synonym 2-Bromo-6-methylphenol
PubChem CID 11095329
IUPAC Name 2-bromo-6-methylphenol
SMILES CC1=C(O)C(Br)=CC=C1

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 187.04
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Bromo-o-cresol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.