accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOFFHHH2O

CAS RN 13319-75-0

IUPAC Name: trifluoroborane dihydrate
Synonyms: boron trifluoride dihydrate
Molecular Weight (g/mol): 103.84
Molecular Formula: BF3H4O2
InChi Key: MJCYPBSRKLJZTB-UHFFFAOYSA-N
SMILES: O.O.FB(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)