Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trifluoride Dihydrate

Catalog No. 359963500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-996-3500ML 500 mL
35-996-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 35-996-3500ML Supplier Sigma Aldrich Supplier No. 359963500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trifluoride dihydrate can be used as: An activator in Pechmann condensation to synthesize 4-methylcoumarin derivatives by the reaction between substituted phenols and methyl acetoacetate. An initiator for the polymerization of benzoxazine.A catalyst in the regioselective Fries rearrangement of phenolic esters to give hydroxyphenyl alkyl/aryl ketones.A catalyst to prepare 8-hydroxymethyltricyclene acetate by the reaction between camphene and formaldehyde in the presence of methylene chloride-acetic anhydride solvent.

Specifications

CAS 13319-75-0
Quantity 500 mL
Molecular Formula BF3 -+ 2H2O
Linear Formula BF3 -+ 2H2O
MDL Number MFCD00149901
Molecular Weight (g/mol) 103.84 g/mol
Refractive Index n20/D 1.316
Density 1.636 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.