accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1332-09-8

IUPAC Name: 2-[9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
Synonyms: 2-[9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-2H,3H,3aH,3bH,4H,5H,10H,11H-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
Molecular Weight (g/mol): 504.6
Molecular Formula: C28H37FO7
InChi Key: CIWBQSYVNNPZIQ-UHFFFAOYNA-N
SMILES: CCC(=O)OCC(=O)C1(OC(=O)CC)C(C)CC2C3CCC4=CC(=O)C=CC4(C)C3(F)C(O)CC12C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Color 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)