accessibility menu, dialog, popup

UserName

Viewing profile as user:
HFFNa

CAS RN 1333-83-1

IUPAC Name: sodium hydrogen difluoride
Synonyms: sodium bifluoride
Molecular Weight (g/mol): 62
Molecular Formula: F2HNa
InChi Key: BFXAWOHHDUIALU-UHFFFAOYSA-M
SMILES: [H+].[F-].[F-].[Na+]
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)