Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bifluoride, Technical, Ricca Chemical
SDP

Catalog No. RDCS0323500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0323500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0323500 Supplier Ricca Chemical Company Supplier No. RDCS0323500B1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1333-83-1
Molecular Formula F2HNa
Molecular Weight (g/mol) 62.00
InChI Key BFXAWOHHDUIALU-UHFFFAOYSA-M
IUPAC Name sodium hydrogen difluoride
SMILES [H+].[F-].[F-].[Na+]

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Identification Pass
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Bifluoride
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.