accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13331-19-6

IUPAC Name: 2-propylbenzene-1,3-diol
Synonyms: 2-propylbenzene-1,3-diol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: XDCMHOFEBFTMNL-UHFFFAOYSA-N
SMILES: CCCC1=C(O)C=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results