Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Propylresorcinol, 98%

Catalog No. AAH2702406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2702406 5 g
AAH2702403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2702406 Supplier Thermo Scientific Chemicals Supplier No. H2702406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13331-19-6 | C9H12O2 | 152.19 g/mol

2-n-propylresorcinol is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-propylresorcinol is a pharmaceutical intermediate.

Solubility
Solightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13331-19-6
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00100568
InChI Key XDCMHOFEBFTMNL-UHFFFAOYSA-N
Synonym 1,3-benzenediol, propyl, 2-propylresorcinol, propyl-1,3-benzenediol, 1,3-benzenediol,2-propyl, 2-propylbenzol-1,3-diol, propyl resorcinol, 2-propyl resorcinol, 2-n-propylresorcinol, pubchem19795, acmc-20ann2
PubChem CID 262060
IUPAC Name 2-propylbenzene-1,3-diol
SMILES CCCC1=C(O)C=CC=C1O

Specifications

Melting Point 101°C to 105°C
Boiling Point 280°C
Flash Point 135°C (275°F)
Odor Phenol-like
Quantity 5 g
Solubility Information Solightly soluble in water.
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-n-Propylresorcinol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.