accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133373-24-7

IUPAC Name: (2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)propanoic acid
Synonyms: (2S)-3-[4-(tert-butoxy)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}propanoic acid
Molecular Weight (g/mol): 473.57
Molecular Formula: C29H31NO5
InChi Key: WTLSDEYZKFJXFT-SANMLTNESA-N
SMILES: CN([C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)