CAS RN 133387-28-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-N,N-diethyl-6-nitroaniline, 96%, Thermo Scientific™
CAS: 133387-28-7 Synonym: (2-Chloro-6-nitrophenyl)diethylamine