CAS RN 133407-86-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MG-115
Potent reversible proteasome inhibitor (Ki = 21 nM and 35 nM for 20S and 26S proteasome, respectively).