Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MG-115

Catalog No. 4747805MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-478-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-478-05MG Supplier MilliporeSigma™ Supplier No. 4747805MG
Add to Cart
Add to Cart

Potent reversible proteasome inhibitor (Ki = 21 nM and 35 nM for 20S and 26S proteasome, respectively).

Peptide Sequence: Z-Leu-Leu-Nva-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by HPLC
Target Proteins 20S proteasome
CAS 133407-86-0
Molecular Formula C25H39N3O5
Formula Weight 461.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.