accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOONHOSS

CAS RN 1334172-66-5

IUPAC Name: 5-(1,2-dithiolan-3-yl)-N-(2,5,8,11-tetraoxatridecan-13-yl)pentanamide
Synonyms: 5-(1,2-dithiolan-3-yl)-N-(2,5,8,11-tetraoxatridecan-13-yl)pentanamide
Molecular Weight (g/mol): 395.57
Molecular Formula: C17H33NO5S2
InChi Key: DVOKZPCLVOHRAR-UHFFFAOYNA-N
SMILES: COCCOCCOCCOCCNC(=O)CCCCC1CCSS1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results