Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-(3,6,9,12-Tetraoxatridecyl)-alpha-lipoamide 90.0+%, TCI America™
SDP

Catalog No. T3199100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3199100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3199100MG Supplier TCI America Supplier No. T3199100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1334172-66-5
Molecular Formula C17H33NO5S2
Molecular Weight (g/mol) 395.573
MDL Number MFCD21363251
InChI Key DVOKZPCLVOHRAR-MRXNPFEDSA-N
Synonym (R)-5-(1,2-Dithiolan-3-yl)-N-(3,6,9,12-tetraoxatridecyl)valeramide, mPEG4-(R)-Lipoamide
PubChem CID 98590430
IUPAC Name 5-[(3R)-dithiolan-3-yl]-N-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES COCCOCCOCCOCCNC(=O)CCCCC1CCSS1

Specifications

Quantity 100 mg
Formula Weight 395.57
Percent Purity ≥90.0% (HPLC)
Physical Form Liquid
Chemical Name or Material (R)-N-(3,6,9,12-Tetraoxatridecyl)-alpha-lipoamide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.