accessibility menu, dialog, popup

UserName

Viewing profile as user:
PdClPNH3CH3CCH3CH3CH3H3CH2CH3CCH3CH3(E)

CAS RN 1334497-06-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 462.35
Molecular Formula: C20H35ClNPPd
InChi Key: ASQSTGRGIHOSGZ-NXZCPFRHSA-M
SMILES: Cl[Pd+].C\C=C\[CH2-].CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results