Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PdCl(crotyl)Amphos

Catalog No. AA4721603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4721603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4721603 Supplier Thermo Scientific Chemicals Supplier No. 04721603
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1334497-06-1 | C20H35ClNPPd | 462.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1334497-06-1
Molecular Formula C20H35ClNPPd
Molecular Weight (g/mol) 462.35
MDL Number MFCD29059853
InChI Key ASQSTGRGIHOSGZ-NXZCPFRHSA-M
Synonym Chloro(crotyl)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II), Crotyl(Amphos)palladium(II) chloride
SMILES Cl[Pd+].C\C=C\[CH2-].CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C

Specifications

Color White
Physical Form Powder
Quantity 1 g
Formula Weight 493.32
Chemical Name or Material PdCl(crotyl)Amphos
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.