accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1335-32-6

IUPAC Name: tris(λ²-lead(2+)) diacetate tetrahydroxide
Synonyms: lead acetate, basic
Molecular Weight (g/mol): 807.7
Molecular Formula: C4H10O8Pb3
InChi Key: VLOJXAQYHIVPFI-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Grade 
  • (1)
  • (1)
  • (4)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)