Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Subacetate, ACS Reagent, Solstice

Catalog No. 60045462 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-462 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-462 Supplier Solstice Supplier No. 237159500G

ACS Reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1335-32-6
MDL Number MFCD00013041

Specifications

Physical Form Solid
Quantity 500 g
Linear Formula Pb(C2H3O2)2 · 2Pb[OH]2
UN Number UN1616
Formula Weight 807.72g/mol
Grade ACS Reagent
Packaging Poly bottle
Chemical Name or Material Lead Subacetate
Hazard Statement P201-P273-P281-P308 + P313-P501
Precautionary Statement H351-H360Df-H373-H410
DOTInformation 6.1/PGIII
EINECSNumber 215-630-3
RTECSNumber OF8750000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.