accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133545-30-9

IUPAC Name: [6'-(diphenylphosphanyl)-2',3',4,4',5,6-hexamethoxy-[1,1'-biphenyl]-2-yl]diphenylphosphane
Synonyms: [6'-(diphenylphosphanyl)-2',3',4,4',5,6-hexamethoxy-[1,1'-biphenyl]-2-yl]diphenylphosphane
Molecular Weight (g/mol): 702.72
Molecular Formula: C42H40O6P2
InChi Key: IYTHLSFAUKKSGJ-UHFFFAOYSA-N
SMILES: COC1=CC(P(C2=CC=CC=C2)C2=CC=CC=C2)=C(C(OC)=C1OC)C1=C(C=C(OC)C(OC)=C1OC)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results