Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ (S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(diphenylphosphine), 97%

Catalog No. AA47270MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47270MD 250 mg
AA4727003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47270MD Supplier Thermo Scientific™ Supplier No. 47270MD
Add to Cart

CAS: 133545-30-9 | C42H40O6P2 | 702.72 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133545-30-9
Molecular Formula C42H40O6P2
Molecular Weight (g/mol) 702.72
MDL Number MFCD30533249
InChI Key IYTHLSFAUKKSGJ-UHFFFAOYSA-N
Synonym (S)-(4,4',5,5',6,6'-Hexamethoxy[1,1'-biphenyl]-2,2'-diyl)bis(diphenylphosphane);
IUPAC Name [6'-(diphenylphosphanyl)-2',3',4,4',5,6-hexamethoxy-[1,1'-biphenyl]-2-yl]diphenylphosphane
SMILES COC1=CC(P(C2=CC=CC=C2)C2=CC=CC=C2)=C(C(OC)=C1OC)C1=C(C=C(OC)C(OC)=C1OC)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 702.71
Percent Purity 97%
Physical Form Solid
ShelfLife 5 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.