accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133550-49-9

IUPAC Name: 2-(2,3-dihydro-1H-indole-1-carbonyl)-3-(3,4-dihydroxyphenyl)prop-2-enenitrile
Synonyms: 2-(2,3-dihydroindole-1-carbonyl)-3-(3,4-dihydroxyphenyl)prop-2-enenitrile
Molecular Weight (g/mol): 306.32
Molecular Formula: C18H14N2O3
InChi Key: MYKCTDWWIWGLHW-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C(C#N)C(=O)N2CCC3=CC=CC=C23)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results